Orders LinkContactUsLinkComponent
Filter sets
Filter sets
Filter set 38, excitation BP 470/40, beam splitter FT 495, emission BP 525/50
Catalog # ZEIS0000001031346
Supplier:   ZEISS
Filter sets
Catalog # ZEIS0000001031346
Supplier:  ZEISS
Filter sets
Filter set 38, excitation BP 470/40, beam splitter FT 495, emission BP 525/50
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Filter set 38, excitation BP 470/40, beam splitter FT 495, emission BP 525/50
  • Seq:
    08