Plugs for test tubes
12 mm, Red
Catalog # 212-0525
Supplier: BOETTGER PAUL
Jump to:
Product Details & Documents
Polyethylene (PE). Ribbed plugs with two sealing ribs, very tight sealing.
- Not autoclavable
- Easy handling with a high, ribbed grip
- Easy to open by tilting 'one-hand operation'
Specifications
- Colour:Red
- Size:12 mm