Plugs for test tubes
12 mm, Blue
212-0524
Polyethylene (PE). Ribbed plugs with two sealing ribs, very tight sealing.
- Not autoclavable
- Easy handling with a high, ribbed grip
- Easy to open by tilting 'one-hand operation'
- Colour:Blue
- Seq:02
- Size:12 mm