Orders LinkContactUsLinkComponent
Slide containers, SlideRite®
Transport containers for slides, SlideRite II, 2
Catalog # 720-2327
Supplier:   CELLPATH
Slide containers, SlideRite®
Catalog # 720-2327
Supplier:  CELLPATH
Slide containers, SlideRite®
Slide containers, SlideRite®
Transport containers for slides, SlideRite II, 2
Price
98,70 €
Log in for availability
1 * 100 items
Product Details & Documents
Slide holders designed for use with one or two slides.
  • Slide- and a click-closed lid
  • Slide stays in the container to apply the swab
Suitable for shipping, storing, or staining slides.
Specifications
  • No. of slides:
    2
  • Description:
    Transport containers for slides, SlideRite II