Orders LinkContactUsLinkComponent
Face shield
Face shield, with helmet adapter, Anti-fog, Cellulose acetate
  111-8231
 :   UVEX
Face shield
  111-8231
 :  UVEX
Face shield
Face shield
Face shield, with helmet adapter, Anti-fog, Cellulose acetate
46,10 €
1 * 1 items
These polycarbonate and cellulose acetate visors can be attached to helmets by using a length adapter and can be locked in three positions.
  • Robust visor with specially developed adapter for attachment to side helmet slots
  • Extremely lightweight plastic design
  • Material thickness: 1 mm
 : Visors are supplied with adapter but without helmet. Visors are compatible with helmets uvex airwing.
  • Type:
    Anti-fog
  • Description:
    Face shield, with helmet adapter
  • Material:
    Cellulose acetate
  • Seq:
    01
Complementary Products
  •  
    Description
    Colour
  • 111-8232
    Safety helmet, Airwing B
    White
    1 * 1 items - 12,70 €
  • 111-8235
    Safety helmet, Airwing B
    Yellow
    1 * 1 items - 12,70 €
  • 111-8216
    Safety helmet, Airwing B
    Orange
    1 * 1 items - 12,70 €
  • 111-8217
    Safety helmet, Airwing B
    Red
    1 * 1 items - 12,70 €
  • 111-8218
    Safety helmet, Airwing B
    Green
    1 * 1 items - 12,70 €
  • 111-8219
    Safety helmet, Airwing B
    Blue
    1 * 1 items - 12,70 €
  • 111-8234
    Safety helmet, Airwing B-WR
    White
    1 * 1 items - 18,40 €
  • 111-8237
    Safety helmet, Airwing B-WR
    Yellow
    1 * 1 items - 18,40 €
  • 111-8253
    Safety helmet, Airwing B-WR
    Orange
    1 * 1 items - 18,40 €
  • 111-8254
    Safety helmet, Airwing B-WR
    Red
    1 * 1 items - 18,40 €
  • 111-8255
    Safety helmet, Airwing B-WR
    Green
    1 * 1 items - 18,40 €
  • 111-8256
    Safety helmet, Airwing B-WR
    Blue
    1 * 1 items - 18,40 €