Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Screw caps for Jerricans

Screw caps for Jerricans

50 mm, 5 and 10 L jerricans
Catalog # 216-1394
Supplier:   Kautex
Screw caps for Jerricans
Catalog # 216-1394
Supplier:  Kautex
Screw caps for Jerricans
Screw caps for Jerricans

50 mm, 5 and 10 L jerricans

Price
3,80 €
Log in for availability
1 * 1 items
Product Details & Documents

Documents

Specifications
  • For:
    5 and 10 L jerricans
  • Thread:
    50 mm
  • Seq:
    14
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.