Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
25 ml, 0,1 ml, 0,045 ± ml
Catalog # BURK9695-1025
Supplier: BURKLE
Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
Catalog # BURK9695-1025
Supplier: BURKLE
Jump to:
Product Details & Documents
Borosilicate glass 3.3, class B.
- Adjusted for Ex
- Blue ring graduations, Schellbach stripe
- Precise dosing using the press button, exact fine titration with the micro-screw and precise discharge jet
Graduation according to Category B (Tolerances according to DIN EN ISO 385).
Certifications: Graduation according to Category B (Tolerances according to DIN EN ISO 385).
Documents
Specifications
- Capacity:25 ml
- Division:0,1 ml
- Tolerance:0,045 ± ml
Accessories
Product Information
- Catalog #AvailabilityPrice
- Spare nozzle, PP
Product Information
- Catalog #AvailabilityPrice
- Cap without feed tube for 1000 ml bottle