Orders LinkContactUsLinkComponent
Precast gels, Reliant™
Precast gels, Reliant™
Reliant™ DNA Gel, 20-well, 2% SeaKem® Gold agarose with TBE and EtBr, 100≥3000 bp
Catalog # 733-1581
Supplier:   LONZA
Precast gels, Reliant™
Catalog # 733-1581
Supplier:  LONZA
Precast gels, Reliant™
Precast gels, Reliant™
Reliant™ DNA Gel, 20-well, 2% SeaKem® Gold agarose with TBE and EtBr, 100≥3000 bp
Price
315,00 €
Log in for availability
1 * 20 items
Product Details & Documents
Reliant™ precast agarose minigels are designed for rapid and reproducible resolution of DNA fragments from 8 bp to 10 kb. Each precast agarose gel is precision cast using our SeaKem® Gold or NuSieve™ 3:1 agarose and are available in a variety of well formats and agarose concentrations.
  • Gels are cast from either SeaKem® or NuSieve® agarose and are certified DNase-free
  • Gels specifically for RNA separation are guaranteed RNase-free
  • Compatible with most minigel chambers
Reliant™ precast agarose minigels are available either pre-stained with ethidium bromide or without stain. The Reliant™ gel System is ideal for analysis of PCR, restriction digest fragments or most cloning needs.
Delivery information: Supplied in boxes of 20 gels.
Specifications
  • Description:
    Reliant™ DNA Gel, 20-well, 2% SeaKem® Gold agarose with TBE and EtBr, 100≥3000 bp
  • Seq:
    12