Orders LinkContactUsLinkComponent
Charts
C210, –18...+37 °C, 7 days
Catalog # 620-1501
Supplier:   DICKSON
Charts
Catalog # 620-1501
Supplier:  DICKSON
Charts
C210, –18...+37 °C, 7 days
Price
79,10 €
Log in for availability
1 * 60 items
Product Details & Documents
Specifications
  • Description:
    C210
  • Seq:
    21
  • Range:
    –18...+37 °C
  • Recording time:
    7 days