QuickBlue RealQuick L. monocytogenes. Complete Real-time PCR kit using the Taqman technique for sensitive and specific detection of the species indicated in food and othe r samples. The kit contains the primer and probe set, Taq polymerase, UNG enzym 1 * 50 Tests
710-3325
Frequently Bought Together