Sample Bags without Closure
600×400 mm, 0,05 mm
Catalog # SCAC10034060
Supplier: SCANCELL FOLIEN VETRIEB
Jump to:
Product Details & Documents
Bags made from PE transparent, for the storage or transport of samples.
Available in several sizes.
Caution: Supplied without closure ties.
Documents
SCAC2046_TDS_EN SCAC20771_TDS_EN
Datasheet
Specifications
- L×W:600×400 mm
- Seq:11
- Thickness:0,05 mm