PCR racks
96-Well PCR rack, Red
Catalog # RATI6114104
Supplier: RATIOLAB
Jump to:
Product Details & Documents
Racks made of PP and are characterised by their very thin but uniform walls.
- 8×12 positions, micro test plate format
- Autoclavable
- Available in different colors for the convenient identification of different types of samples
- Transparent lids available in two different heights (Lids must be ordered seperately)
Work and storage rack for 0,2 ml and 0,5 ml PCR tubes, PCR strips, and PCR segments.
Specifications
- Colour:Red
- Description:96-Well PCR rack
- Seq:04
Accessories
Product Information
- Catalog #AvailabilityPrice
- Lid, transparent, height 14 mm
Product Information
- Catalog #AvailabilityPrice
- Lid, transparent, height 28 mm