6-Hydroxyindole 97%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Indol-6-ol, 6-Indolol, IMEXINE OBA, 6-Hydroxy-1H-indole, 1H-Indol-6-ol
- CAS Number:2380-86-1
- Formula:C₈H₇NO
- MDL Number:MFCD00152101
- Molecular Weight:133.15 g/mol
- Melting Pt:124…130 °C
- Storage Temperature:Refrigerator
- EINECS:417-020-4
- UN:3077
- ADR:9,III
Documents
ACRO37901
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Glistening crystalline powder (May darken on storage)
- Infrared spectrum:Conforms
- Melting point:124 to 130 °C
- HPLC:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



