3-Chlorophenol 99%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:108-43-0
- Formula:C₆H₅ClO
- MDL Number:MFCD00002256
- Molecular Weight:128.56 g/mol
- Merck Index:13,02173
- Boiling Pt:214 °C (760 mmHg)
- Melting Pt:33…35 °C
- Density:1.218 g/cm³
- Flash Pt:97 °C
- Storage Temperature:Ambient
- EINECS:203-582-6
- UN:2020
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:Clear yellow to brown Liquid after melting
- Infrared spectrum:Conforms
- Refractive index:1.5650 to 1.5670 (20°C, 589 nm)
- GC:≥98.5 %
- Iron (Fe):≤50 ppm
- Magnesium (Mg):≤100 ppm
- Potassium (K):≤50 ppm
- Sodium (Na):≤100 ppm
- Silicon dioxide (SiO2):≤1000 ppm
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



