Orders LinkContactUsLinkComponent
Dip slides
Dip slides
Plate Count/VRBG, 10
Catalog # OXOIGFD06A
Dip slides
Catalog # OXOIGFD06A
CAS Number:  
Dip slides
Plate Count/VRBG, 10
CAS Number:  
Price
30,90 €
Log in for availability
1 * 10 items
Product Details & Documents
Simple and safe for an unskilled operator to use, these dip slides are coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective. This sterile dip slide can be inoculated in the ward or doctor’s surgery and then returned to its plastic container for easy transport back to the laboratory.
  • Make colony counting for the bacterial assessment simple
Documents
Specifications
Specifications
  • Pk:
    10
  • Description:
    Plate Count/VRBG
Specifications