ContactUsLinkComponent
G-CAPSULE is a single use electroelution device for rapid recovery of proteins and nucleic acids from electrophoresis gel. Isolates 20bp of DNA or RNA or protein 4kDa in 60 seconds. Simply pick up the protein band with the Gel-Pick, assemble with Gen 1 * 55 items
Catalog # 786-001
Supplier:  G-Biosciences
undefined
G-CAPSULE is a single use electroelution device for rapid recovery of proteins and nucleic acids from electrophoresis gel. Isolates 20bp of DNA or RNA or protein 4kDa in 60 seconds. Simply pick up the protein band with the Gel-Pick, assemble with Gen 1 * 55 items
Catalog # 786-001
Supplier:  G-Biosciences