Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Swingsette®, M515
Tissue processing/embedding cassettes, Swingsette®, M515
Blue
Catalog # 720-1638
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M515
Catalog # 720-1638
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Swingsette®, M515
Tissue processing/embedding cassettes, Swingsette®, M515
Blue
Price
258,00 €
Log in for availability
1 * 1.500 items
Product Details & Documents
Acetal polymer.
  • Integral, hinged lid, with large tab for easy opening
  • Disposable plastic cassettes highly resistant to histological solvents
  • Flow-through slots maximise fluid exchange
  • Front face inclined 45° to enable use with leading automatic labelling systems.
Packaging: Each case contains three boxes of 500 cassettes.
Specifications
  • Colour:
    Blue