Orders LinkContactUsLinkComponent
Assortment Boxes
Assortment Boxes
330×220×70 mm
Catalog # SEMA0430
Supplier:   SEMADENI
Assortment Boxes
Catalog # SEMA0430
Supplier:  SEMADENI
Assortment Boxes
Assortment Boxes
330×220×70 mm
Price
46,40 €
Log in for availability
1 * 1 items
Product Details & Documents
Assortment and storage boxes, made of polystyrene, are crystal clear with hinged lid and without divisions.
 : The partitions allow storage boxes (330×220 mm) to be divided into convenience in many variants. Comes in different colours. The lids are transparent in colour.
Specifications
  • Seq:
    03
  • W×D×H:
    330×220×70 mm
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Lids, PS, transparent, for compartments, 108×54 mm
    Lids, PS, transparent, for compartments, 108×54 mm
    SEMA0446
    Log in for availability
    1 * 1 items - 3,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Lids, PS, transparent, for compartments, 54×54 mm
    Lids, PS, transparent, for compartments, 54×54 mm
    SEMA0445
    Log in for availability
    1 * 1 items - 2,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Compartments, PS for storage boxes, yellow, 108×54 mm, 45 mm height
    Compartments, PS for storage boxes, yellow, 108×54 mm, 45 mm height
    SEMA0436
    Log in for availability
    1 * 1 items - 2,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Compartments, PS for storage boxes, red, 54×54 mm, 45 mm height
    Compartments, PS for storage boxes, red, 54×54 mm, 45 mm height
    SEMA0435
    Log in for availability
    1 * 1 items - 2,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Compartments, PS for storage boxes, blue, 108×108 mm, 45 mm height
    Compartments, PS for storage boxes, blue, 108×108 mm, 45 mm height
    SEMA0437
    Log in for availability
    1 * 1 items - 4,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Lids, PS, transparent, for compartments, 162×108 mm
    Lids, PS, transparent, for compartments, 162×108 mm
    SEMA0448
    Log in for availability
    1 * 1 items - 5,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Lids, PS, transparent, for compartments, 215×108 mm
    Lids, PS, transparent, for compartments, 215×108 mm
    SEMA0449
    Log in for availability
    1 * 1 items - 5,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Compartments, PS for storage boxes, green, 162×108 mm, 45 mm height
    Compartments, PS for storage boxes, green, 162×108 mm, 45 mm height
    211-2743
    Log in for availability
    1 * 1 items - 7,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Compartments, PS for storage boxes, grey, 215×108 mm, 45 mm height
    Compartments, PS for storage boxes, grey, 215×108 mm, 45 mm height
    SEMA0439
    Log in for availability
    1 * 1 items - 8,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Lids, PS, transparent, for compartments, 108×108 mm
    Lids, PS, transparent, for compartments, 108×108 mm
    SEMA0447
    Log in for availability
    1 * 1 items - 4,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Compartments, PS for storage boxes, grey, 322×108 mm, 45 mm height
    Compartments, PS for storage boxes, grey, 322×108 mm, 45 mm height
    SEMA0440
    Log in for availability
    1 * 1 items - 9,80 €