Orders LinkContactUsLinkComponent
Dressing jars with lid
Dressing jars with lid
124 mm, 130 mm
Catalog # BOCH8622
Supplier:   BOCHEM
Dressing jars with lid
Catalog # BOCH8622
Supplier:  BOCHEM
Dressing jars with lid
Dressing jars with lid
124 mm, 130 mm
Price
44,70 €
Log in for availability
1 * 1 items
Product Details & Documents
Jar made of stainless steel.
  • Stackable
  • Lid with handle
  • Very good chemical and temperature resistance
Specifications
  • Ext.Ø:
    124 mm
  • Height:
    130 mm