Orders LinkContactUsLinkComponent
Microscope slides, UniMark®
Microscope slides, UniMark®
Slides in standard packs, Green
Catalog # 631-1205
Supplier:   MARIENFELD
Microscope slides, UniMark®
Catalog # 631-1205
Supplier:  MARIENFELD
Microscope slides, UniMark®
Microscope slides, UniMark®
Slides in standard packs, Green
Price
3,70 €
Log in for availability
1 * 50 items
Product Details & Documents
The bright printed colours of UniMark® slides frosted ends contrast well with the markings.
  • Frosted end for labelling
  • Soda-lime glass, Hydrolytic Class 3
  • Different colours to identify preparations securely
  • Thin coating to facilitate the processing of slides in automated systems
W×D×H: 76×26×1 mm
For in vitro diagnostic (IVD) applications.
Documents
Certificates
Search by Lot Number
Specifications
  • Colour:
    Green
  • Description:
    Slides in standard packs
  • Seq:
    15