Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CANNON CALIBRATIEOLIE N14 1 * 500 mL
Catalog # CANN9727-C34
Supplier:
CANNON INSTRUMENT
Expand image
CANNON CALIBRATIEOLIE N14 1 * 500 mL
Catalog # CANN9727-C34
Supplier:
CANNON INSTRUMENT
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals