ContactUsLinkComponent
Thermometer, graduation 1.0c, tolerance 2.0c, range -10:210c, mercury free - filled with a low toxicity blue spirit, nominal le ngth 305mm with item 30/210/10 length 405mm, reinforced bulb for stirring, permanent amber stain graduations. 1 * 10 items
Catalog # BIBB30/210/10
Supplier:  SciLabware
Thermometer, graduation 1.0c, tolerance 2.0c, range -10:210c, mercury free - filled with a low toxicity blue spirit, nominal le ngth 305mm with item 30/210/10 length 405mm, reinforced bulb for stirring, permanent amber stain graduations. 1 * 10 items
Catalog # BIBB30/210/10
Supplier:  SciLabware
Frequently Bought Together