Orders LinkContactUsLinkComponent
Filter sets
Filter sets
Filter set 45, excitation BP 560/40, beam splitter FT 585, emission BP 630/75
Catalog # ZEIS4880459100400
Supplier:   ZEISS
Filter sets
Catalog # ZEIS4880459100400
Supplier:  ZEISS
Filter sets
Filter set 45, excitation BP 560/40, beam splitter FT 585, emission BP 630/75
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Filter set 45, excitation BP 560/40, beam splitter FT 585, emission BP 630/75
  • Seq:
    12