Orders LinkContactUsLinkComponent
Filter pump
Filter pump
Water pump 5720, with non return valve, female connection G 3/8 to water top, outside Ø: 16, 6 mm (230 g)
Catalog # USBE5720
Supplier:   USBECK Laborgeräte
Filter pump
Catalog # USBE5720
Filter pump
Filter pump
Water pump 5720, with non return valve, female connection G 3/8 to water top, outside Ø: 16, 6 mm (230 g)
Price
80,40 €
Log in for availability
1 * 1 items
Product Details & Documents
Water jet pump in nickel-plated brass.
  • Stable and solid
  • A side knob allows connection to a vacuum pipe with an interior diameter of 8 mm
  • Available with or without non-return valve
  • Supplied with water connection for pipes with an internal diameter of 14 mm or bolting for connection to a tap
  • Pressure gauge available as an option for all models
Documents
Specifications
  • Description:
    Water pump 5720, with non return valve, female connection G 3/8 to water top, outside Ø: 16, 6 mm (230 g)
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Water pump manometer
    Water pump manometer
    181-0121
    Log in for availability
    1 * 1 items - 66,60 €
Product Information
  • Catalog #
    Availability
    Price
  • G 1/2 connection for water pump
    G 1/2 connection for water pump
    USBE5741
    Log in for availability
    1 * 1 items - 22,50 €