Orders LinkContactUsLinkComponent
Face shields
Face shields
Anti-UV (UV 380), Cellulose acetate, 195×412 mm
Catalog # 111-1577
Supplier:   UVEX
Face shields
Catalog # 111-1577
Supplier:  UVEX
Face shields
Face shields
Anti-UV (UV 380), Cellulose acetate, 195×412 mm
Price
40,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Robust visor with integrated forehead cover, clear cellulose acetate lens and additional protective cap made from impact-resistant PS.
  • Twist ratchet enables simple and reliable adjustment to any head size
  • Flip-up mechanism allows the visor to be set in any position
  • Easy lens replacement
 : EN 166
Documents
Specifications
  • Type:
    Anti-UV (UV 380)
  • L×W:
    195×412 mm
  • Material:
    Cellulose acetate
Accessories
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Replacement visor lens
    Replacement visor lens
    111-2543
    Cellulose acetate
    Log in for availability
    1 * 1 items - 13,70 €
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Replacement visor lens, anti-fog
    Replacement visor lens, anti-fog
    111-8229
    Cellulose acetate
    Log in for availability
    1 * 1 items - 30,70 €