Volumetric flasks, class A
20 ml, 10/19, 0,04 ± ml
Catalog # POULRB258/WAC/D/5
Supplier: Poulten & Graf
Jump to:
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Documents
POULRB258^WAC^J^2_TDS_EN
Datasheet
Specifications
- NS:10/19
- Capacity:20 ml
- Seq:04
- Tolerance:0,04 ± ml
- Cat. No.:POULRB258/WAC/D/5