Tip boxes, dispolab®
Box for refill filter tips in rack with transparent snap-on lid, Grey
Catalog # KART9756
Supplier: KARTELL
Jump to:
Product Details & Documents
Empty boxes for use with refill tips.
- Semitransparent boxes in PC have a horizontal sliding cover
- Yellow boxes in PP feature a snap-on cover
Specifications
- Colour:Grey
- Description:Box for refill filter tips in rack with transparent snap-on lid