Orders LinkContactUsLinkComponent
Filter discs
4, 20 mm
Catalog # LENZ07180420
Supplier:   Lenz Laborglas
Filter discs
Catalog # LENZ07180420
Supplier:  Lenz Laborglas
Filter discs
Filter discs
4, 20 mm
Price
44,70 €
Log in for availability
1 * 10 items
Product Details & Documents
Glass filter discs centred with unfused rim.
Specifications
  • Ø:
    20 mm
  • Porosity:
    4
  • Seq:
    26