Orders LinkContactUsLinkComponent
Filter discs
Filter discs
0, 30 mm
Catalog # LENZ07180030
Supplier:   Lenz Laborglas
Filter discs
Catalog # LENZ07180030
Supplier:  Lenz Laborglas
Filter discs
Filter discs
0, 30 mm
Price
44,70 €
Log in for availability
1 * 10 items
Product Details & Documents
Glass filter discs centred with unfused rim.
Documents
Specifications
  • Ø:
    30 mm
  • Porosity:
    0
  • Seq:
    03