Orders LinkContactUsLinkComponent
Tissue grinders, Potter type
Tissue grinders, Potter type
30 ml, 25×175 mm
Catalog # KART6130
Supplier:   KARTELL
Tissue grinders, Potter type
Catalog # KART6130
Supplier:  KARTELL
Tissue grinders, Potter type
Tissue grinders, Potter type
30 ml, 25×175 mm
Price
145,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Int.Ø×H:
    25×175 mm
  • Capacity:
    30 ml
  • Seq:
    05