Sedimentation cones, Imhoff type
Imhoff cones, sclae up to 100 ml, ring mark at 1000 ml, with stopcock
Catalog # LENZ05670005
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
Made of borosilicate glass 3.3.
- White graduations
- With or without stopcock
- Excellent chemical and temperature resistance
Specifications
- Type:with stopcock
- Description:Imhoff cones, sclae up to 100 ml, ring mark at 1000 ml
- Cat. No.:LENZ05670005
- Graduation:0 - 2 ml
- Division:0,1 ml