Orders LinkContactUsLinkComponent
Dissolved oxygen bottles
Oxygen bottles acc. to Winkler, 100-150 ml, 14/23
Catalog # 528-1205
Supplier:   Lenz Laborglas
Dissolved oxygen bottles
Catalog # 528-1205
Supplier:  Lenz Laborglas
Dissolved oxygen bottles
Dissolved oxygen bottles
Oxygen bottles acc. to Winkler, 100-150 ml, 14/23
Price
32,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Glass, clear.
  • White inscription label
  • Oblique ground NS stopper
  • Optional stopper clamp is available
For the determination of dissolved oxygen in water according to Winkler. The capacity is engraved on the bottle with an accuracy of ± 0,01 ml. Each bottle is individually adjusted with its stopper (non interchangeable). Each stopper and bottle is identified with the same serial number.
Specifications
  • Capacity:
    100-150 ml
  • Description:
    Oxygen bottles acc. to Winkler
  • NS stopper:
    14/23
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Clamps NS 14/23 for Dissolved Oxygen Bottles
    Clamps NS 14/23 for Dissolved Oxygen Bottles
    705-0564
    Log in for availability
    1 * 1 items - 7,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Clamps NS 19/26 for Dissolved Oxygen Bottles
    Clamps NS 19/26 for Dissolved Oxygen Bottles
    705-0565
    Log in for availability
    1 * 1 items - 8,20 €