Orders LinkContactUsLinkComponent
Extractor thimbles for solids
Extractor thimbles for solids
250 ml, 60/46 NS
Catalog # LENZ05431149
Supplier:   Lenz Laborglas
Extractor thimbles for solids
Catalog # LENZ05431149
Supplier:  Lenz Laborglas
Extractor thimbles for solids
Extractor thimbles for solids
250 ml, 60/46 NS
Price
159,00 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For glass inserts for solids.
Documents
Specifications
  • Extraction cap.:
    250 ml
  • Socket size:
    60/46 NS
  • Seq:
    03