Orders LinkContactUsLinkComponent
Raschig rings
Raschig rings
Raschig ring, 3 mm, 3 mm
Catalog # LENZ05127003
Supplier:   Lenz Laborglas
Raschig rings
Catalog # LENZ05127003
Supplier:  Lenz Laborglas
Raschig rings
Raschig rings
Raschig ring, 3 mm, 3 mm
Price
190,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Soda lime glass.
  • For packing columns
  • Good chemical resistance
  • Good thermal resistance
Specifications
  • Ext.Ø:
    3 mm
  • Description:
    Raschig ring
  • Height:
    3 mm
  • Seq:
    01