Orders LinkContactUsLinkComponent
Multi-way stopcocks, Discofix®
Two-way stopcocks, Blue
Catalog # 720-2579
Supplier:   B. Braun
Multi-way stopcocks, Discofix®
Catalog # 720-2579
Supplier:  B. Braun
Multi-way stopcocks, Discofix®
Multi-way stopcocks, Discofix®
Two-way stopcocks, Blue
Price
145,00 €
Log in for availability
1 * 50 items
Product Details & Documents
Suitable for all pressure infusion systems <2 bar.
  • Continuous flow channels for accurate pressure monitoring
  • Swivel lock can be moved axially or radially for fast and secure connection
  • PVC and latex-free
Suitable for all pressure infusion systems <2 bar.
Specifications
  • Colour:
    Blue
  • Description:
    Two-way stopcocks