Orders LinkContactUsLinkComponent
Poly-spoons
Poly-spoons
One broad spoon end, one spatula end, 400 mm
Catalog # HAMMHSN176-40
Supplier:   HAMMACHER KARL
Poly-spoons
Catalog # HAMMHSN176-40
Supplier:  HAMMACHER KARL
Poly-spoons
Poly-spoons
One broad spoon end, one spatula end, 400 mm
Price
40,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Poly-spoons made of stainless-steel.
  • Double-ended: One spoon and one spatula end
Specifications
  • Description:
    One broad spoon end, one spatula end
  • Length:
    400 mm
  • Seq:
    08