Orders LinkContactUsLinkComponent
Uridine ≥99%
Uridine ≥99%
Glass bottle for solids, 100 g
Catalog # A15227.22
Uridine ≥99%
Catalog # A15227.22
CAS Number:  58-96-8
Uridine ≥99%
Uridine ≥99%
Glass bottle for solids, 100 g
CAS Number:  58-96-8
Price
491,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Purity: 99% CAS No.: 58-96-8 Molecular Formula: C9H12N2O6 Formula Weight: 244.20 Melting Point: 166-169° Beilstein Reference: 754904 Merck Reference: 14,9877 Optical Rotation: +9.5° (c=2 in water) MDL No.: MFCD00006526
Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.20 g/mol
  • Merck Index:14,9877
  • Melting Pt:166…169 °C
  • Storage Temperature:Ambient
  • EINECS:200-407-5
Specifications
Specifications
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g