Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

3-Chlorophenol 98%

Glass bottle for solids, 250 g
Catalog # A13671.30
Irritant
Environment
3-Chlorophenol 98%
Catalog # A13671.30
CAS Number:  108-43-0
3-Chlorophenol 98%
3-Chlorophenol 98%

Glass bottle for solids, 250 g

CAS Number:  108-43-0
Price
313,00 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 98+% CAS No.: 108-43-0 Molecular Formula: C6H5ClO Formula Weight: 128.56 Flash Point: 120°(248°F) Refractive Index: 1.5660 Melting Point: 30-34° Boiling Point: 214° Beilstein Reference: 1634401 Merck Reference: 14,2154 UN No.: 2020 MDL No.: MFCD00002256

  • CAS Number:108-43-0
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002256
  • Molecular Weight:128.56 g/mol
  • Merck Index:14,2154
  • Boiling Pt:214 °C
  • Melting Pt:30…34 °C
  • Density:1.245
  • Flash Pt:120 °C (248 °F)
  • Storage Temperature:Ambient
  • EINECS:203-582-6
  • UN:2020
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications