Orders LinkContactUsLinkComponent
3-Chlorophenol 98%
Glass bottle for solids, 50 g
  A13671.18
3-Chlorophenol 98%
  A13671.18
 :  108-43-0
3-Chlorophenol 98%
3-Chlorophenol 98%
Glass bottle for solids, 50 g
 :  108-43-0
84,70 €
1 * 50 g
Purity: 98+% CAS No.: 108-43-0 Molecular Formula: C6H5ClO Formula Weight: 128.56 Flash Point: 120°(248°F) Refractive Index: 1.5660 Melting Point: 30-34° Boiling Point: 214° Beilstein Reference: 1634401 Merck Reference: 14,2154 UN No.: 2020 MDL No.: MFCD00002256
  •  :108-43-0
  •  :C₆H₅ClO
  •  :MFCD00002256
  •  :128.56 g/mol
  •  :14,2154
  •  :214 °C
  •  :30…34 °C
  •  :1.245
  •  :120 °C (248 °F)
  •  :Ambient
  •  :203-582-6
  •  :2020
  •  :6.1,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g