N,N-Dimethylformamide ≥99%
Glass bottle, 250 mL
Purity: 99% CAS No.: 68-12-2 Molecular Formula: HCON(CH3)2 Formula Weight: 73.09 Flash Point: 57°(135°F) Refractive Index: 1.4310 Odour: Amine-like pH: 7 Melting Point: -61° Boiling Point: 153° Beilstein Reference: 605365 Merck Reference: 14,3243 UN No.: 2265 MDL No.: MFCD00003284
: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
- :68-12-2
- :HCON(CH₃)₂
- :MFCD00003284
- :73.09 g/mol
- :14,3243
- :153 °C
- :–61 °C
- :0.944
- :57 °C (135 °F)
- :Ambient
- :200-679-5
- :2265
- :3,III
- :01-2119475605-32
Documents
- Pack type:Glass bottle
- Pk:250 mL