Orders LinkContactUsLinkComponent
N,N-Dimethylformamide ≥99%
N,N-Dimethylformamide ≥99%
Glass bottle, 250 mL
  A13547.AK
N,N-Dimethylformamide ≥99%
  A13547.AK
 :  68-12-2
N,N-Dimethylformamide ≥99%
N,N-Dimethylformamide ≥99%
Glass bottle, 250 mL
 :  68-12-2
30,50 €
1 * 250 mL
Purity: 99% CAS No.: 68-12-2 Molecular Formula: HCON(CH3)2 Formula Weight: 73.09 Flash Point: 57°(135°F) Refractive Index: 1.4310 Odour: Amine-like pH: 7 Melting Point: -61° Boiling Point: 153° Beilstein Reference: 605365 Merck Reference: 14,3243 UN No.: 2265 MDL No.: MFCD00003284
 :  Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
  •  :68-12-2
  •  :HCON(CH₃)₂
  •  :MFCD00003284
  •  :73.09 g/mol
  •  :14,3243
  •  :153 °C
  •  :–61 °C
  •  :0.944
  •  :57 °C (135 °F)
  •  :Ambient
  •  :200-679-5
  •  :2265
  •  :3,III
  •  :01-2119475605-32
  • Pack type:
    Glass bottle
  • Pk:
    250 mL
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.