Order Entry
Slovenia
Orders LinkContactUsLinkComponent
N,N-Dimethylformamide ≥99%

N,N-Dimethylformamide ≥99%

Glass bottle, 2.500 mL
Catalog # A13547.0F
Health
Flammable
Irritant
N,N-Dimethylformamide ≥99%
Catalog # A13547.0F
CAS Number:  68-12-2
N,N-Dimethylformamide ≥99%
N,N-Dimethylformamide ≥99%

Glass bottle, 2.500 mL

CAS Number:  68-12-2
Price
85,60 €
Log in for availability
1 * 2.500 mL
Product Details & Documents

Purity: 99% CAS No.: 68-12-2 Molecular Formula: HCON(CH3)2 Formula Weight: 73.09 Flash Point: 57°(135°F) Refractive Index: 1.4310 Odour: Amine-like pH: 7 Melting Point: -61° Boiling Point: 153° Beilstein Reference: 605365 Merck Reference: 14,3243 UN No.: 2265 MDL No.: MFCD00003284

Synonyms:  Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
  • CAS Number:68-12-2
  • Formula:HCON(CH₃)₂
  • MDL Number:MFCD00003284
  • Molecular Weight:73.09 g/mol
  • Merck Index:14,3243
  • Boiling Pt:153 °C
  • Melting Pt:–61 °C
  • Density:0.944
  • Flash Pt:57 °C (135 °F)
  • Storage Temperature:Ambient
  • EINECS:200-679-5
  • UN:2265
  • ADR:3,III
  • REACH:01-2119475605-32
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.