Orders LinkContactUsLinkComponent
D-(-)-Aspartic acid ≥99%
D-(-)-Aspartic acid ≥99%
Glass bottle for solids, 5 g
  B21184.06
D-(-)-Aspartic acid ≥99%
  B21184.06
 :  1783-96-6
D-(-)-Aspartic acid ≥99%
D-(-)-Aspartic acid ≥99%
Glass bottle for solids, 5 g
 :  1783-96-6
32,60 €
1 * 5 g
Purity: 99% CAS No.: 1783-96-6 Molecular Formula: C4H7NO4 Formula Weight: 133.10 Melting Point: 300° dec. Beilstein Reference: 1723529 Merck Reference: 14,840 Optical Rotation: -25° (c=1 in 1N HCl) MDL No.: MFCD00063081
 :  D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH
  •  :1783-96-6
  •  :C₄H₇NO₄
  •  :MFCD00063081
  •  :133.1 g/mol
  •  :14,840
  •  :>300 °C (dec.)
  •  :1.66
  •  :Ambient
  •  :217-234-6
  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g