D-(-)-Aspartic acid ≥99%
Glass bottle for solids, 5 g
Purity: 99% CAS No.: 1783-96-6 Molecular Formula: C4H7NO4 Formula Weight: 133.10 Melting Point: 300° dec. Beilstein Reference: 1723529 Merck Reference: 14,840 Optical Rotation: -25° (c=1 in 1N HCl) MDL No.: MFCD00063081
: D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH
- :1783-96-6
- :C₄H₇NO₄
- :MFCD00063081
- :133.1 g/mol
- :14,840
- :>300 °C (dec.)
- :1.66
- :Ambient
- :217-234-6
Documents
- Pack type:Glass bottle for solids
- Pk:5 g