2,4,6-Trichlorophenol ≥98%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
2,4,6-Trichlorophenol ≥98%
Catalog # A11681.22
Supplier: Thermo Fisher Scientific
CAS Number: 88-06-2
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 88-06-2 Molecular Formula: C6H3Cl3O Formula Weight: 197.45 Flash Point: 99°(210°F) Physical Form: Cryst solid Density: 1.675 Appearance: White to pale brown Odour: Phenol-like Melting Point: 63-68° Boiling Point: 245-246° Beilstein Reference: 776729 Merck Reference: 14,9644 UN No.: 2020 MDL No.: MFCD00002172
- CAS Number:88-06-2
- Formula:C₆H₃Cl₃O
- MDL Number:MFCD00002172
- Molecular Weight:197.45 g/mol
- Merck Index:14,9644
- Boiling Pt:245…246 °C
- Melting Pt:63…68 °C
- Density:1.675
- Flash Pt:99 °C (210 °F)
- Storage Temperature:Ambient
- EINECS:201-795-9
- UN:2020
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



