Microtubes, with Thread
Micro tubes without screw cap, without skirted base, 0,5 ml, 5000/case, 0,5 ml
Catalog # SARS72.733.010
Supplier: SARSTEDT
Jump to:
Product Details & Documents
PP, translucent.
- Safe screw cap with O-ring
- Sterile or non sterile
- Autoclavable
For transport, storage and sample processing in the laboratory.
Documents
Specifications
- Packed:5000/case
- Recommended working volume:0,5 ml
- Capacity:0,5 ml
- Description:Micro tubes without screw cap, without skirted base
- Seq:9