Orders LinkContactUsLinkComponent
Microtubes
Tubes with screw cap non assembled, with writing space, 0,5 ml, 5000/case, 0,5 ml
  SARS72.730.107
 :   SARSTEDT
Microtubes
  SARS72.730.107
 :  SARSTEDT
Microtubes
Tubes with screw cap non assembled, with writing space, 0,5 ml, 5000/case, 0,5 ml
670,00 €
1 * 5.000 items
PP, translucent
  • Excellent chemical resistance
  • Excellent temperature resistance
Without or with skirted base.
  • Packed:
    5000/case
  • Recommended working volume:
    0,5 ml
  • Capacity:
    0,5 ml
  • Description:
    Tubes with screw cap non assembled, with writing space
Accessories
  •  
    Description
  • SARS65.713.006
    Colour coded inserts, blue
    1 * 1.000 items - 14,70 €
  •  
    Description
  • SARS65.712
    Screw cap with seperate retaining loop, neutral
    1 * 5.000 items - 218,00 €
  •  
    Description
  • SARS65.713
    Colour coded inserts, neutral
    1 * 5.000 items - 66,50 €
  •  
    Description
  • SARS65.713.002
    Colour coded inserts, red
    1 * 5.000 items - 72,80 €
  •  
    Description
  • SARS65.713.007
    Colour coded inserts, violet
    1 * 1.000 items - 13,30 €
  •  
    Description
  • SARS65.713.009
    Colour coded inserts, white
    1 * 1.000 items - 14,70 €
  •  
    Description
  • SARS65.713.010
    Colour coded inserts, black
    1 * 5.000 items - 66,50 €
  •  
    Description
  • SARS65.713.004
    Colour coded inserts, yellow
    1 * 5.000 items - 72,80 €
  •  
    Description
  • SARS65.713.005
    Colour coded inserts, green
    1 * 5.000 items - 85,30 €