Pipette tips
Type D, 10 µl, +, Transparent, Eppendorf Reference, Research and Research Plus, 96/box, 480/inner case 1920/case
Catalog # SARS70.1115.200
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
These high quality tips are suitable for the most common pipettes.
- Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
- Available loose in bags, packed in racks or in Tip StackPack reload systems
Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.
Specifications
- Packed:96/box, 480/inner case 1920/case
- Sterile:+
- Volume:10 µl
- Seq:11
- Tip style:Type D
- Tip colour:Transparent
- For use with:Eppendorf Reference, Research and Research Plus