Orders LinkContactUsLinkComponent
Microtubes, Prepared
Microtubes, Prepared
Microtubes, prepared, coagulation citrate, with attached push cap closure, 1,0 ml, 1000/case
Catalog # SARS41.1506
Supplier:   SARSTEDT
Microtubes, Prepared
Catalog # SARS41.1506
Supplier:  SARSTEDT
Microtubes, Prepared
Microtubes, prepared, coagulation citrate, with attached push cap closure, 1,0 ml, 1000/case
Price
188,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
PP, translucent with PP screw cap.
  • Type of preparation: Trisodium citrate solution
  • With paper label
  • Graduated
  • Recommended centrifugation: 1800×g
With screw cap or push-on cap.
Specifications
  • Packed:
    1000/case
  • Capacity:
    1,0 ml
  • Description:
    Microtubes, prepared, coagulation citrate, with attached push cap closure
  • Seq:
    13