Transfer pipettes
1,0 ml, 17 - 22 µl, 104, - ml, -, Bulk
SARS86.1180
: SARSTEDT
LDPE, disposable.
- No risk of injury or infection from broken glass
- Disposable: No carry over or cross contamination
- Can be heat-sealed for sample storage and transport
- Environmentally neutral, ensuring non-toxic disposal
- Packed:Bulk
- Sterile:-
- Capacity:1,0 ml
- Length:104
- Seq:02
- Drop vol.:17 - 22 µl
- Graduation:- ml