Orders LinkContactUsLinkComponent
Microtubes
Tubes with screw cap assembled, sterile, 0,5 ml, 1000/case, 0,5 ml
Catalog # SARS72.730.105
Supplier:   SARSTEDT
Microtubes
Catalog # SARS72.730.105
Supplier:  SARSTEDT
Microtubes
Tubes with screw cap assembled, sterile, 0,5 ml, 1000/case, 0,5 ml
Price
167,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
PP, translucent
  • Excellent chemical resistance
  • Excellent temperature resistance
Without or with skirted base.
Specifications
  • Packed:
    1000/case
  • Recommended working volume:
    0,5 ml
  • Capacity:
    0,5 ml
  • Description:
    Tubes with screw cap assembled, sterile
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Colour coded inserts, blue
    Colour coded inserts, blue
    SARS65.713.006
    Log in for availability
    1 * 1.000 items - 14,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Screw cap with seperate retaining loop, neutral
    Screw cap with seperate retaining loop, neutral
    SARS65.712
    Log in for availability
    1 * 5.000 items - 218,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Colour coded inserts, neutral
    Colour coded inserts, neutral
    SARS65.713
    Log in for availability
    1 * 5.000 items - 66,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Colour coded inserts, red
    Colour coded inserts, red
    SARS65.713.002
    Log in for availability
    1 * 5.000 items - 72,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Colour coded inserts, violet
    Colour coded inserts, violet
    SARS65.713.007
    Log in for availability
    1 * 1.000 items - 13,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Colour coded inserts, white
    Colour coded inserts, white
    SARS65.713.009
    Log in for availability
    1 * 1.000 items - 14,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Colour coded inserts, black
    Colour coded inserts, black
    SARS65.713.010
    Log in for availability
    1 * 5.000 items - 66,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Colour coded inserts, yellow
    Colour coded inserts, yellow
    SARS65.713.004
    Log in for availability
    1 * 5.000 items - 72,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Colour coded inserts, green
    Colour coded inserts, green
    SARS65.713.005
    Log in for availability
    1 * 5.000 items - 85,30 €