Microtubes, with Thread
Micro tubes with writing space and graduation, without screw cap, with skirted base, 2,0 ml, 5000/case
SARS72.609.001
: SARSTEDT
PP, translucent.
- Safe screw cap with O-ring
- Sterile or non sterile
- Autoclavable
For transport, storage and sample processing in the laboratory.
Documents
- Packed:5000/case
- Capacity:2,0 ml
- Description:Micro tubes with writing space and graduation, without screw cap, with skirted base
- Seq:26